Eisco® Linear Air Track Systems | Avantor
Eisco® Linear Air Track Systems
Supplier:   Wards
Eisco® Linear Air Track Systems
Supplier:  Wards
Certificates
Eisco® Linear Air Track Systems
Eisco® Linear Air Track Systems
3 Options Available Below
Product Details & Documents
These kits provide a cost-effective and friction-free system for the study of dynamics.
  • Cost-effective study of dynamics
  • Friction-free environment
  • Effective in the field of collisions
  • Comprehensive accessory pack
  • Other topics include velocity, kinetic energy, and more
Linear air tracks are particularly effective in the field of collisions where conservation of momentum is dramatically demonstrated.
These kits contain everything required for a vast array of experiments covering velocity, force and acceleration, potential energy, kinetic energy, conservation of energy, SHM, and motion on level and inclined planes. Gliders are designed to accept standard slotted masses for additional loads. The kits are also supplied with a comprehensive accessory pack to facilitate every type of interaction.
Accessories information: For measurement of glider’s speed, standard light gates and timers are required. Except where noted, the kits do not contain an air blower.
Ordering information: These kits include: (1) linear air track and (1) accessory pack, which comes with a range of accessories including vehicles with mass attachments, catapults, and buffers.
Product Family Options
Informations sur le produit
  • Désignation
    Availability
    Price
  • 1.5 m linear air track system
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - 1 195,50 $
    1 195,50 $
    Each
       1 195,50 $
    La quantité minimale de commande est de 1
    Supplier Number: PH0362A
  • 2.0 m Linear air track system
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - 1 556,20 $
    1 556,20 $
    Each
       1 556,20 $
    La quantité minimale de commande est de 1
    Supplier Number: PH0362
el 4
Informations sur le produit
  • Désignation
    Availability
    Price
  • 2.0 m linear air track system, with quiet air source
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - 3 016,05 $
    3 016,05 $
    Each
       3 016,05 $
    Supplier Number: PH0362SET
Accessories
Informations sur le produit
  • Catalog #
    Availability
    Price
  • Accessories set for linear air track
    Accessories set for linear air track
    470224-380
    Log in pour la disponibilité
    Each - $264.50