Transformer Accessory Kit | Avantor
Transformer Accessory Kit
Transformer Accessory Kit
Catalog # 470157-638
Supplier:   Wards
Fournisseur #:  PH0849AK2
Transformer Accessory Kit
Catalog # 470157-638
Supplier:  Wards
Transformer Accessory Kit
Transformer Accessory Kit
Transformer Accessory Kit
Liquidation
53,75 $
Vente
Valable jusqu'au 31/12/2026
Log in pour la disponibilité
Each
Product Details & Documents
Demonstrate Lenz's Law!
  • Excellent accessory to a variable gap magnet
Lenz’s law transformer accessory kit to demonstrate the production of large magnetic fields and lenz’s law damping on pendulums with and without slits.
Spécifications
  • Désignation:
    Transformer Accessory Kit