ContactUsLinkComponent
Bio-Plex® 200 System Validation and Calibration Tools, Bio-Rad
Bio-Plex® 200 System Validation and Calibration Tools, Bio-Rad
  100445-794
 :  Bio-Rad
Bio-Plex® 200 System Validation and Calibration Tools, Bio-Rad
  100445-794
 :  Bio-Rad
 :  171203050
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 
  • Description:
    Bio-Plex® Reservoir, pkg 1
  • Cat. no.:
    100445-794
  • Contents:
    For use with Bio-Plex Manager™ 5.0 Software
  • Supplier No.:
    171203050
System performance validation and calibration tools for the Bio-Plex® 200 System ensure reliability and consistency of assay results
The Bio-Plex® Calibration Kit standardizes signal output from day to day and from instrument to instrument. The Bio-Plex® System allows calibration at two different settings; broad range standards provides the widest available dynamic range, while the calibration for low range standards with Bio-Plex Manager™ Software StatLIA curve fitting provides the greatest sensitivity.
Perform routine instrument maintenance, calibration, and validation procedures. Assess and document the performance of all system components. Discriminate between instrument and assay problems. Automated validation with both high and low PMT settings. Broadened classification regions from region 1 to 100. Bio-Plex® Flashlight for visual inspection and adjustment of sample-needle height. Two needle-height adjustment options to accommodate various plate types. Bio-Plex® Reservoir to enable unattended operation of maintenance functions.
Both validation and calibration protocols are automated by Bio-Plex Manager™ Software, and records are stored in a secure Bio-Plex® database file. Procedures are performed using a maintenance, calibration, and validation (MCV) plate that holds the bead sets and solutions. The Bio-Plex® Validation Kit 4.0 includes bead sets for validation routines (optics, reporter, classify, and fluidics validation) using Bio-Plex Manager™ Software and the Bio-Plex® MCV plate IV.