Flask Tongs
Flask Tongs
Catalog # 470165-458
Supplier: Wards
Fournisseur #: CH0739
Jump to:
Product Details & Documents
With cork-lined jaws, these stainless steel flask tongs are resilient and long lasting.
- Cork-Lined Jaws
- Stainless Steel Construction
- Tongs Fit Flasks of Any Shape from 100 to 2000 mL
Length: 30 cm
Spécifications
- Désignation:Flask Tongs