Organic Micro Glassware Set | Avantor
Organic Micro Glassware Set
Organic Micro Glassware Set
Catalog # 470165-440
Supplier:   Wards
Fournisseur #:  CH0881G
Organic Micro Glassware Set
Catalog # 470165-440
Supplier:  Wards
Organic Micro Glassware Set
Organic Micro Glassware Set
Organic Micro Glassware Set
Price
$456.00
Log in pour la disponibilité
Each
Product Details & Documents
The Perfect Set for Organic Chemistry Labs!
Ideal for organic chemistry labs, enabling applications in distillation, fractional distillation, separation, purification, and synthesis. This set consists of 8 items of 14/23 ground joint.
Spécifications
  • Désignation:
    Organic Micro Glassware Set