CleanFoam®, Sterile Swabs, Texwipe®
Fournisseur: ITW Texwipe
Certificats
À propos de cet article
The sterile swabs are constructed from the highest quality 100% polyurethane foam. Complete thermal bond construction eliminates adhesive contamination.
- Gamma irradiated
- Certificates of irradiation and compliance attached to each case
- Packaging labeled for global use
- Pre sterilized swabs eliminate repackaging, documentation and validation of autoclaved swabs
- Lot code and expiration date clearly marked on each package
The swabs are made to exacting and consistent tolerances using high precision automated processes. The easy to open, individually wrapped swabs ensure sterility at point-of-use. The swabs are visible through clear packaging which also offers visibility of seal integrity.
Ordering information: Packaged in a reclosable, silicone free bag.
Frequently Bought Together