Premium Bosshead
Premium Bosshead
Catalog # 470165-424
Supplier: Wards
Fournisseur #: PRBH02
Jump to:
Product Details & Documents
Great for Connecting Rods and Apparatus in Tight Places without Sacrificing Strength
- Threaded metal one-piece clamping screws
- Able to hold rods up to 15 mm in diameter
- Holds rods orthogonal (90°) to one another
- All metal construction to carry heavy loads and prevent torque damage
Spécifications
- Désignation:Premium Bosshead