Premium Three-Finger Clamp, Cork Lined | Avantor
Premium Three-Finger Clamp, Cork Lined
Premium Three-Finger Clamp, Cork Lined
Catalog # 470165-418
Supplier:   Wards
Fournisseur #:  PRCMP09
Premium Three-Finger Clamp, Cork Lined
Catalog # 470165-418
Supplier:  Wards
Premium Three-Finger Clamp, Cork Lined
Premium Three-Finger Clamp, Cork Lined
Premium Three-Finger Clamp, Cork Lined
Price
$50.00
Log in pour la disponibilité
Each
Product Details & Documents
This three-finger clamp is made of powder-coated steel.
  • Maximum Clamp Opening: 8.5 cm
  • Connecting Rod: 14 cm
  • Powder-Coated Steel Construction
  • Cork-Lined
  • Thread-Fitted Actuating Joints
The clamp is cork-lined and attached to a rugged, aluminum rod.
It has thread-fitted actuating joints and a 10 mm-diameter connecting rod.
Spécifications
  • Désignation:
    Premium Three-Finger Clamp, Cork Lined