- Description:Elements & the Periodic Table Curriculum Learning Module
- Cat. no.:470163-830
- Supplier No.:74-6923
A Great Classroom Solution.
- Includes detailed, standards-based coverage of:
- Introduction to Elements
- Atomic Structure
- Metals
- Metalloids
- Nonmetals
- The Periodic Table
- Groups on the Periodic Table
- Flame Test - Identifying Elements
NewPath’s Elements and the Periodic Table Curriculum Learning Module provides a complete set of visual, digital and hands-on resources designed to support teacher instruction and provide students with the skills to master key concepts on Elements and the Periodic Table.
Each module includes:
1 Flip Chart Set with 10 double-side, laminated charts including a graphic overview and “write-on/wipe-off” activities
1 Student Learning Guide featuring self-directed readings, illustrated explanations, assessment questions and a lab investigation
1 Vocabulary Review Card Set
1 Curriculum Mastery Game (4 players)
1 Interactive Multimedia Lesson containing animated tutorials, interactive activities, visual resources, illustrated narrated glossary, virtual labs, and assessments.