Nylon Burette Brushes
Nylon Burette Brushes
Supplier:   Wards
Nylon Burette Brushes
Supplier:  Wards
Certificates
Nylon Burette Brushes
Nylon Burette Brushes
3 Options Available Below
Product Details & Documents
These brushes are available in a variety of sizes.
  • Durable
  • Long-lasting nylon bristles
These durable brushes will keep your burettes and labware clean.
Product Family Options
Product Information
  • Désignation
    Bristle Material
    Ø tête de la brosse
    Handle Material
    Longueur totale
    Availability
    Price
  • Nylon burette brush, for 50 mL burettes
    Nylon
    19 mm
    Stainless steel
    760 mm
    Log in for availability
    Log in for availability
    Each
    Each - 7,40 $
    7,40 $
    Each
       7,40 $
    Supplier Number: CH0217E
  • Nylon burette brush, for 100 mL burettes
    Nylon
    19 mm
    Stainless steel
    915 mm
    Log in for availability
    Log in for availability
    Each
    Each - 6,70 $
    6,70 $
    Each
       6,70 $
    Supplier Number: CH0217F
  • Nylon burette brush, for 25 mL burettes
    Nylon
    16 mm
    Stainless steel
    610 mm
    Log in for availability
    Log in for availability
    Each
    Each - 5,05 $
    5,05 $
    Each
       5,05 $
    Supplier Number: CH0217D