ContactUsLinkComponent
HRP and AP Antibody Conjugates, Bio-Rad
HRP and AP Antibody Conjugates, Bio-Rad
Catalog # 101645-262
Supplier:  Bio-Rad
CAS Number:  
HRP and AP Antibody Conjugates, Bio-Rad
Catalog # 101645-262
Supplier:  Bio-Rad
Supplier Number:  1706515
CAS Number:  
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 
Specifications
  • Description:
    Goat Anti-Rabbit IgG (H + L)-HRP Conjugate, Blotting Grade
  • Size:
    2 mL
  • Cat. no.:
    101645-262
  • Supplier No.:
    1706515
Specifications
About this item
Bio-Rad Horseradish Peroxidase (HRP) and Alkaline Phosphatase (AP) Secondary Antibody Conjugates are engineered for use with blot detection protocols including colorimetric and chemiluminescent.
Double affinity-purified blotting-grade antibodies are isolated by affinity chromatography, and further purified by cross-adsorption against an unrelated species to eliminate nonspecific immunoglobulins.
High titer blotting-grade antibody conjugates increase assay sensitivity; greater working dilutions (1:3,000) decrease background and increase the signal-to-noise ratio.