Amersham ECL Western Blotting Start Detection Reagents, Cytiva
Fournisseur: Cytiva
Certificats
À propos de cet article
Chemiluminescence detection reagent for detection of high to medium abundant proteins.
- For outine and confirmatory western blotting, high expressed proteins and tagged proteins
- Detection with X-ray film or CCD Imager
- Stable until the expiration date printed on box when stored under recommended conditions
- Signal duration < 3 h
Recommended detection with Amersham Hyperfilm™ (X-ray film) or ImageQuant™ LAS 500. Optimal membrane amersham protran.
Spécifications
- Storage temperature:4...8 °C