Cooling Bath Circulators, CC-Range, Huber
À propos de cet article
Refrigeration bath circulators are suitable for controlling the temperature of externally connected applications and thermoregulation of objects directly in the thermostat bath
The new multi-touch controller Pilot ONE offers a brilliant 5.7" touchscreen with graphic functions and a comfortable navigation menu in 11 languages. All important operating parameters and temperature values are neatly displayed on the touchscreen. Thanks to the new favourites menu, the one-click operation and the integrated technical glossary the operation is very easy, just like on a smartphone. Integrated USB and Ethernet ports allows connection to a PC or network, e.g. for remote control or data transmission.
CC-range has the following advantages and functions: Pilot ONE controller with Plug and Play technology, Precise temperature control, 5.7“ touchscreen and comfortable menu navigation, Overtemperature and low level protection, Safety class III/FL (DIN 12876), Powerful pressure/suction pump, Programmer with calendar and clock functions, 5-point calibration, RS232 interface and Pt100 connection.
Typical applications are: photometers, refractometers, viscometers, double-jacketed reaction vessels and autoclaves. The units can be used in mini-plants and kilo laboratories, for determining the freezing point, for deep temperature calibration, for petroleum tests, for temperature control of measuring devices and experimental setups as well as for materials testing and quality control. Depending on the model, the equipment is able to meet the highest demands; this is ensured by the professional range of functions available on the Pilot ONE controller, such as a colour TFT display, programmer, Pt100 external probe connection, calibration, calendar/clock functions, autostart, RS232 interface as well as extensive safety and warning functions. One further plus point is the electronic upgrade for function extensions using a unit specific e-grade code. This activates additional functions such as the programmer, TAC cascade controlling, temperature ramps, user menus, calendar start, graphic display and process control. The units are made from high quality stainless steel.
Ordering information: Depending on the model, cooling bath circulator could be supplied with a mini-USB cable #54949, bath cover, Adapter nom. dia. 12, dummy plugs, sleeve nuts thread M16x1. Please contact your Avantor representative for standard delivery specification.
Caution: Note: Suffix w denotes water cooled models, wl denotes air/water cooled models.
1 Options Available Below
Product Details & Documents
Refrigeration bath circulators are suitable for controlling the temperature of externally connected applications and thermoregulation of objects directly in the thermostat bath
The new multi-touch controller Pilot ONE offers a brilliant 5.7" touchscreen with graphic functions and a comfortable navigation menu in 11 languages. All important operating parameters and temperature values are neatly displayed on the touchscreen. Thanks to the new favourites menu, the one-click operation and the integrated technical glossary the operation is very easy, just like on a smartphone. Integrated USB and Ethernet ports allows connection to a PC or network, e.g. for remote control or data transmission.
CC-range has the following advantages and functions: Pilot ONE controller with Plug and Play technology, Precise temperature control, 5.7“ touchscreen and comfortable menu navigation, Overtemperature and low level protection, Safety class III/FL (DIN 12876), Powerful pressure/suction pump, Programmer with calendar and clock functions, 5-point calibration, RS232 interface and Pt100 connection.
Typical applications are: photometers, refractometers, viscometers, double-jacketed reaction vessels and autoclaves. The units can be used in mini-plants and kilo laboratories, for determining the freezing point, for deep temperature calibration, for petroleum tests, for temperature control of measuring devices and experimental setups as well as for materials testing and quality control. Depending on the model, the equipment is able to meet the highest demands; this is ensured by the professional range of functions available on the Pilot ONE controller, such as a colour TFT display, programmer, Pt100 external probe connection, calibration, calendar/clock functions, autostart, RS232 interface as well as extensive safety and warning functions. One further plus point is the electronic upgrade for function extensions using a unit specific e-grade code. This activates additional functions such as the programmer, TAC cascade controlling, temperature ramps, user menus, calendar start, graphic display and process control. The units are made from high quality stainless steel.
Ordering information: Depending on the model, cooling bath circulator could be supplied with a mini-USB cable #54949, bath cover, Adapter nom. dia. 12, dummy plugs, sleeve nuts thread M16x1. Please contact your Avantor representative for standard delivery specification.
Caution: Note: Suffix w denotes water cooled models, wl denotes air/water cooled models.
Specifications for Product Family
Spécifications
- Catalog No:
- 10221-712
- 10221-714
- 10221-720
- 10221-894
- 10221-716
- 10221-718
- 10221-896
- 10220-836
- 10220-838
- 10221-090
- 10221-092
- 10221-094
- 10221-096
- 10221-098
- 10221-100
- 10221-722
- 10221-724
- 10221-726
- 10220-780
- 10220-782
- 10220-784
- 10220-786
- 10221-018
- 10221-022
- 10221-024
- 10221-030
- 10221-036
- 10221-042
- Environmentally Preferable:
- Safer Human and Environmental Health
- Safer Human and Environmental Health
- Safer Human and Environmental Health
- Safer Human and Environmental Health
- Safer Human and Environmental Health
- Safer Human and Environmental Health
Spécifications
Product Family Options
Air cooled models
Informations sur le produit
- ModèlePlage de températureFor Use WithW×D×HBath CapacityPuissance cryoHeating CapacityPump PressurePump SuctionWeightAvailabilityPrice
- CC-405–40...+200 °C208 V, 2~ 60 Hz14.57×18.11×26.73"1.32 gal.0.7 kW at 0 °C; 0.7 kW at 20 °C; 0.03 kW at −40 °C;1.6 kW8.72 gpm (10.15 psi) max.5.81 gpm ( 5.8 psi) max.121.28 lbs.Discontinued-Supplier Number: -Spécifications:
More
Specifications
Cat. No.10221-712Environmentally Preferable - CC-410–45...+200 °C208 V, 2~ 60 Hz16.54×22.24×28.31"5.81 gal.0.8 kW at 0 °C; 0.8 kW at 20 °C; 0.1 kW at −40 °C;2.45 kW8.72 gpm (10.15 psi) max.5.81 gpm (5.8 psi) max.158.76 lbs.Discontinued-Supplier Number: -Spécifications:
More
Specifications
Cat. No.10221-720Environmentally Preferable - CC-415–40...+200 °C208 V, 2~ 60 Hz16.14×18.9×30.08"1.32 gal.1 kW at 0 °C; 1.2 kW at 20 °C; 0.05 kW at −40 °C;1.6 kW8.72 gpm (10.15 psi) max.5.81 gpm (5.8 psi) max.132.3 lbs.Discontinued-Supplier Number: -Spécifications:
More
Specifications
Cat. No.10221-716Environmentally Preferable - CC-505–50...+200 °C208 V, 2~ 60 Hz16.14×18.9×30.08"1.32 gal.1 kW at 0 °C; 1.2 kW at 20 °C; 0.15 kW at −40 °C;1.6 kW8.72 gpm (10.15 psi) max.5.81 gpm (5.8 psi) max.132.3 lbs.Discontinued-Supplier Number: -Spécifications:
More
Specifications
Cat. No.10221-896Environmentally Preferable - CC-510–50...+200 °C208 V, 2~ 60 Hz23.82×27.8×44.72"6.87 gal.2.1 kW at 0 °C; 2.1 kW at 20 °C; 0.4 kW at −40 °C;1.65 kW8.19 gpm (8.7 psi) max.6.34 gpm (5.07 psi) max.216.09 lbs.Discontinued-Supplier Number: -Spécifications:
More
Specifications
Cat. No.10220-838Environmentally Preferable - CC-515–55...+200 °C460 V, 3~ 60 Hz23.82×27.8×44.72"6.87 gal.3.3 kW at 0 °C; 3.3 kW at 20 °C; 0.6 kW at −40 °C;3.6 kW8.19 gpm (8.7 psi) max.6.34 gpm (5.07 psi) max.216.09 lbs.Discontinued-Supplier Number: -Spécifications:
More
Specifications
Cat. No.10221-092Environmentally Preferable - CC-515–55...+200 °C208 V, 3~ 60 Hz23.82×27.8×44.72"6.87 gal.3.3 kW at 0 °C; 3.3 kW at 20 °C; 0.6 kW at −40 °C;1.5 kW8.19 gpm (8.7 psi) max.6.34 gpm (5.07 psi) max.313.11 lbs.Discontinued-Supplier Number: -Spécifications:
More
Specifications
Cat. No.10221-094Environmentally Preferable - CC-805–80...+100 °C208 V, 2~ 60 Hz16.14×18.9×30.08"1.32 gal.0.5 kW at 0 °C; 0.5 kW at 100 °C; 0.3 kW at −60 °C;1.5 kW8.72 gpm (10.15 psi) max.5.81 gpm (5.8 psi) max.176.4 lbs.Discontinued-Supplier Number: -Spécifications:
More
Specifications
Cat. No.10221-722Environmentally Preferable - CC-820–80...+100 °C208 V, 3~ 60 Hz21.22×24.76×43.39"4.49 gal.1.2 kW at 0 °C; 1.2 kW at 100 °C; 0.6 kW at −60 °C;3.6 kW8.19 gpm (8.7 psi) max.6.34 gpm (5.07 psi) max.584.33 lbs.Discontinued-Supplier Number: -Spécifications:
More
Specifications
Cat. No.10221-724Environmentally Preferable - CC-902–90...+200 °C460 V, 3~ 60 Hz21.65×23.62×35.87"1.32 gal.1.2 kW at 0 °C; 1.2 kW at 100 °C; 0.6 kW at −60 °C;1.8 kW8.72 gpm (10.15 psi) max.5.81 gpm (5.8 psi) max.306.5 lbs.Discontinued-Supplier Number: -Spécifications:
More
Specifications
Cat. No.10220-780Environmentally Preferable - CC-905–90...+200 °C460 V, 3~ 60 Hz23.82×27.8×44.72"6.87 gal.2.0 kW at 0 °C; 2.0 kW at 100 °C; 1.0 kW at −60 °C;3 kW8.19 gpm (8.7 psi) max.6.34 gpm (5.07 psi) max.357.21 lbs.Discontinued-Supplier Number: -Spécifications:
More
Specifications
Cat. No.10220-782Environmentally Preferable - CC-K6–25...+200 °C110 V, 1~ 50/60 Hz8.27×15.75×21.5"1.19 gal.0.15 kW at 0 °C; 0.20 kW at 20 °C; 0.05 kW at −20 °C;1.5 kW7.13 gpm (10.15 psi) max.6.6 gpm (5.8 psi) max.55.12 lbs.Discontinued-Supplier Number: -Spécifications:
Safer Human and Environmental Health More
Specifications
Cat. No.10221-018Environmentally PreferableSafer Human and Environmental HealthSafer Human and Environmental Health - CC-K6s–25...+200 °C110 V, 1~ 50/60 Hz8.27×15.75×21.5"1.19 gal.0.21 kW at 0 °C; 0.26 kW at 20 °C; 0.05 kW at −20 °C;1.5 kW7.13 gpm (10.15 psi) max.6.6 gpm (5.8 psi) max.55.12 lbs.Discontinued-Supplier Number: -Spécifications:
Safer Human and Environmental Health More
Specifications
Cat. No.10221-022Environmentally PreferableSafer Human and Environmental HealthSafer Human and Environmental Health - CC-K12–20...+200 °C115 V, 1~ 60 Hz13.78×22.05×16.93"3.17 gal.0.2 kW at 0 °C; 0.25 kW at 20 °C; 0.05 kW at −20 °C;1.5 kW7.13 gpm (10.15 psi) max.6.6 gpm (5.8 psi) max.61.74 lbs.Discontinued-Supplier Number: -Spécifications:
Safer Human and Environmental Health More
Specifications
Cat. No.10221-024Environmentally PreferableSafer Human and Environmental HealthSafer Human and Environmental Health - CC-K15–20...+200 °C115 V, 1~ 60 Hz13.78×22.05×16.93"3.96 gal.0.2 kW at 0 °C; 0.25 kW at 20 °C; 0.05 kW at −20 °C;1.5 kW7.13 gpm (10.15 psi) max.6.6 gpm (5.8 psi) max.61.74 lbs.Discontinued-Supplier Number: -Spécifications:
Safer Human and Environmental Health More
Specifications
Cat. No.10221-030Environmentally PreferableSafer Human and Environmental HealthSafer Human and Environmental Health - CC-K20–30...+200 °C115 V, 1~ 60 Hz13.78×21.85×24.21"5.28 gal.0.35 kW at 0 °C; 0.4 kW at 20 °C; 0.16 kW at −20 °C;1.5 kW7.13 gpm (10.15 psi) max.6.6 gpm (5.8 psi), max.79.38 lbs.Log in pour la disponibilitéEach$28,239.80Supplier Number: 2011.0019.01Spécifications:
Safer Human and Environmental Health More
Specifications
Cat. No.10221-036Environmentally PreferableSafer Human and Environmental HealthSafer Human and Environmental Health - CC-K25–30...+200 °C115 V, 1~ 60 Hz13.78×21.85×24.21"6.6 gal.0.35 kW at 0 °C; 0.4 kW at 20 °C; 0.16 kW at −20 °C;1.5 kW7.13 gpm (10.15 psi) max.6.6 gpm (5.8 psi) max.79.38 lbs.Discontinued-Supplier Number: -Spécifications:
Safer Human and Environmental Health More
Specifications
Cat. No.10221-042Environmentally PreferableSafer Human and Environmental HealthSafer Human and Environmental Health