ContactUsLinkComponent
Mole In The Bag
Catalogue # 470013-628
Fournisseur:  Avantor
undefined
Mole In The Bag
Catalogue # 470013-628
Fournisseur:  Avantor
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
  • • Issued document with your organization's Federal Tax ID Number
  • • Government issued document with your organization's Resale Tax ID Number
  • • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
Spécifications
  • Description:
    Mole in the Bag Inquiry Activity
  • No. of Lab Groups:
    1
  • Cat. no.:
    470013-628
  • Supplier No.:
    WARD470013-628
Spécifications
À propos de cet article
How Big Is A Mole? Why does a mole of copper weigh more than a mole of sulfur?
  • No Additional Equipment Needed
  • Use year after year
Students will answer these inquiry questions and more while introducing the concept of the mole and its importance in chemistry with this visual demonstration. The bags contain several elements and compounds and are available for students to examine, weigh, and explore. Use the mini bags to demonstration fractions of a mole, then bring the bags out again to demonstrate the relationship between moles and molar solutions. This is a great teaching aid for introducing moles, Avogadro's Number, gram molecular weight, and morality.