Flame Test Station Kit | Avantor
Flame Test Station Kit
Flame Test Lab Station Kit, Instructions, 100
Catalog # 470013-550
Supplier:   Avantor
Flame Test Station Kit
Catalog # 470013-550
Supplier:  Avantor
Flame Test Station Kit
Flame Test Station Kit
Flame Test Lab Station Kit, Instructions, 100
Price
$114.85
Log in pour la disponibilité
Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Setting Up Your Flame Test Lab Just Got Easier.
  • Kit contains all necessary materials for lab set up
  • Further experiments can be performed with spectroscopes, available separately
  • Solutions include: calcium nitrate, copper(II) nitrate, potassium nitrate, sodium nitrate, and strontium nitrate
This kit has everything you need, conveniently arranged and clearly explained. Simply open up the five included bottles of solutions, Lay out the wood splints, pass out the instructions and you are on your way to a successful lab activity. Cobalt glass is also included to help distinguish between the potassium and the sodium flame test.
Spécifications
  • Désignation:
    Flame Test Lab Station Kit
  • Includes:
    Instructions
  • No. of Uses:
    100