ART®, Filter Tips, Thermo Scientific
Manufactured using a special process, tips are furnished with a unique ART® filter to eliminate any risk of cross-contamination between samples, which is particularly important in molecular biology. They are suitable for applications involving the use of DNA or RNA, as well as radioactive and infectious samples.
- Certified free of RNase, DNase, DNA and pyrogens
- Sterile tips: Sterilised by gamma radiation
- 'Low Retention' tips: Extremely hydrophobic surface for lower retention, superior precision and reproducibility of measurements
: Supplied loose bagged and non sterile, sterile in racks, in BreakThru packaging (recyclable, economical packaging in a box), or individually wrapped.
- Catalog No:
- 72830-562
- 72830-556
- Environmentally Preferable:
- Low Manufacturing Impact
- Low Manufacturing Impact
- Environmental certification:
- ISO14001 (Certificate Registration No. EMS 752603)
- ISO14001 (Certificate Registration No. EMS 752603)
Frequently Bought Together