Matrix™ Blank and Alphanumeric Storage Tubes, Thermo Scientific
Catalogue # CA10366-080
Fournisseur: Thermo Fisher Scientific
Spécifications
- Sterility:Non Sterile
- Capacity:1.4 ml
- Description:Alphanumeric storage tube
- Material:PP
- Cat. no.:CA10366-080
- Packaging:10 Latch Racks of 96 tubes
- Supplier No.:4252-11
Spécifications
À propos de cet article
Storage tubes ideal for storing valuable samples where 2D tracking is not required.
- Ideal for scaling up from microplate or deep well block sample storage
Visual identification of samples can be made using the alphanumeric code on the bottom of each tube, and the tubes are compatible with the full line of seals and capping equipment.