Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Description:Gravimetric Analysis
- No. of Lab Groups:15
- Cat. no.:470163-120
- Includes:Teacher's Guide and Student Copymaster
- Supplier No.:IS8028
Specifications
About this item
Determine the Unknown!
- Classic AP Chemistry Experiment
- Instructions Included
Determine the identity of a metal carbonate by gravimetric analysis. Gravimetric analysis is a technique through which the amount of an analyte (the ion being analyzed) can be determined through the measurement of mass. You will complete the analyses comparing the masses of two compounds containing the analyte. The formula weight and the identity of the unknown are determined using gravimetric analysis.
Includes:
Calcium Chloride, anhydrous
Silver Nitrate