ContactUsLinkComponent
Determination of the Empirical Formula of Magnesium Oxide
Catalog # 470163-114
Supplier:  Ward's Science
undefined
Determination of the Empirical Formula of Magnesium Oxide
Catalog # 470163-114
Supplier:  Ward's Science
Supplier Number:  IS8025
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 
Specifications
  • Description:
    Determination of the Empirical Formula of Magnesium Oxide
  • No. of Lab Groups:
    15
  • Cat. no.:
    470163-114
  • Supplier No.:
    IS8025
Specifications
About this item
Quantitative Stoichiometry
  • Classic AP Chemistry Experiment
  • Instructions Included
The quantitative stoichiometric relationships governing mass and amount will be studied using the combustion reaction of magnesium metal. Magnesium is reacted with oxygen from the air in a crucible, and the mass before and after the oxidation is measured. The resulting masses are used to calculate the experimental empirical formula of magnesium oxide, which is then compared to the theoretical empirical formula. A crucible and Bunsen burner will be used to heat magnesium metal to burning. This lab illustrates the (i) law of conservation of mass and (ii) the law of constant composition.
Includes:
Magnesium Metal