ContactUsLinkComponent
Determination of the Properties of Buffer Solutions
Determination of the Properties of Buffer Solutions
Catalog # 470163-112
Supplier:  Ward's Science
undefined
Determination of the Properties of Buffer Solutions
Catalog # 470163-112
Supplier:  Ward's Science
Supplier Number:  IS8024
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 
Specifications
  • Description:
    Determination of the Properties of Buffer Solutions
  • No. of Lab Groups:
    15
  • Cat. no.:
    470163-112
  • Includes:
    Teacher's Guide and Student Copymaster
  • Supplier No.:
    IS8024
Specifications
About this item
What Does A Buffer Do?
  • Classic AP Chemistry Experiment
  • Instructions Included
In this experiment, you will prepare three buffer solutions having different pH values and show that the pH of these solutions does not change significantly when small amounts of acids and bases are added. You will also show that when the same amounts of acids and bases are added to water and to a non-buffer solution (e.g. NaCl solution), the pH changes are large.
Includes:
Sodium Acetate, trihydrate
Ammonium Chloride
Sodium Bicarbonate
Sodium Chloride
Acetic Acid EZ-Prep
Ammonium Hydroxide EZ-Prep
Sodium Hydroxide Solution
Hydrochloric Acid Solution