Bernoulli Bag
Bernoulli Bag
Bernoulli Bag, 4
Catalog # 470163-924
Supplier:   American Scientific
Supplier #:  7040-00
Bernoulli Bag
Catalog # 470163-924
Bernoulli Bag
Bernoulli Bag
Bernoulli Bag, 4
Price
26,40 $
Log in pour la disponibilité
Pack of 4
Product Details & Documents
Explore Bernoulli's principle of air pressure.
  • Comes with a teacher's guide
By placing the bag firmly to your mouth (like a balloon), it will take many breaths to blow it up. But if you hold it out in front of your mouth and blow, air pressure in the stream you produce is reduced, entrapping surrounding air to join in filling up the bag. So you can blow it up with a single breath! This is especially effective after your students have counted many of their own breaths in attempting to fill up the bag!
Spécifications
  • Désignation:
    Bernoulli Bag
  • No. of Parts:
    4