Vacuum Lifter
Vacuum Lifter, 10.5 x 10.5"
Catalog # 470163-920
Supplier: American Scientific
Fournisseur #: AR-510
Jump to:
Product Details & Documents
Demonstrates Atmospheric Pressure.
- Explore additional concepts such as surface area, force, vacuums, and pressure
This durable, thick rubber mat has a simple design. Pick up an ordinary flat lab stool or make a challenge to lift off a table, which is nearly impossible! Show how suction cups stay stuck because of the pushing force of air, not from a pulling "suction" force. Comes with instructions.
Spécifications
- Désignation:Vacuum Lifter
- Dimensions:10.5 x 10.5"