Air Puck Hovercraft
Air puck hovercraft, 7½", 2¼"
Catalog # 470163-906
Supplier: American Scientific
Fournisseur #: 3569-00
Jump to:
Product Details & Documents
Demonstrate Newton’s Laws and the conservation of energy and momentum with the amazing new Air Puck!
- Teacher's guide included, with NGSS alignment
- Requires 4 × AA batteries, not included
The Air Puck hovercraft is perfectly balanced and glides on a cushion of air for nearly frictionless motion across any smooth surface. Sturdy bumpers will survive collisions with walls, shoes, or other Air Pucks. Use it to demonstrate Newton's First Law, uniform linear motion. Or, collide two Air Pucks to demonstrate conservation of momentum in two dimensions!
: Mini hovercraft launcher is recommended but not included.
Spécifications
- Désignation:Air puck hovercraft
- Hauteur:2¼"
- Diameter:7½"