Crime Scene Investigation Lab Activity
Catalog # 470163-328
Supplier: Ward's Science
Specifications
- Description:Crime Scene Investigation Lab
- Cat. no.:470163-328
- Supplier No.:IS9011
Specifications
About this item
Combine Your Forensic Chemistry Knowledge To Solve the Crime!
- Three activities in one convenient kit
- All required chemicals are included
Use your forensic techniques to solve the crime of the missing frogs from the biology classroom. Four possible suspects have been identified by the authorities. Use fingerprints, hair examination, and chemical analysis of ink by thin-layer chromatography to help determine the most likely culprit.
Ordering information: Includes Fingerprinting Powder, Fingerprinting Brushes, Acetate Sheets, Ink Pad, Hand Magnifiers, Forceps, Felt Tip Markers, Chromatography Solvent, Silica Gel Chromatography Sheets, Capillary Tubes, Evidence Envelopes, Microscope Slides, Coverslips, and Teacher's Guide and Student Copymaster.
Delivery: Due to the nature of this item, it cannot be shipped by air under any circumstances. Please select ground shipping at time of checkout.