VWR® Graduated Cylinders, Calibrated To Deliver, Class B
Fournisseur: VWR International
Certificats
À propos de cet article
Cylinder sizes from 25mL to 2000mL feature a hexagonal base and yellow bumper.
- Borosilicate glass fabrication
- Calibrated at 20°C to deliver
“TD” appears on each cylinder and indicates the cylinder is calibrated to deliver
Made from ASTM E438 Type 1 Class A borosilicate 3.3 Glass. As per ASTM Specification E1272 Class B requirements.
Durable white color scale printing for long life in laboratory environment.
Spécifications
- Catalog No:
- 10545-968
- 10545-970
- 10546-054
- 10546-052
- 10546-058
- 10546-056
- 10546-062
- 10546-060
- Form:
- Tall
- Tall
- Tall
- Tall
- Tall
- Tall
- Tall
- Tall