Human Recombinant SCF (from E. coli)
À propos de cet article
Stem cell factor (SCF) is a cytokine made by fibroblasts and endothelial cells. SCF binds to the receptor c-Kit/CD117 and plays a critical role in the maintenance, survival, and differentiation of hematopoietic stem cells. While human SCF shows no activity on murine cells, murine and rat SCF are active on human cells.
- High Quality
- Low Endotoxin
- Affordable
A COA is supplied with each product shipped detailing purity, formulation, biological activity, endotoxin levels, reconstitution instructions and recommended storage conditions. This product is for research use only and is not for use in humans.
Caution: For research use only.
4 Options Available Below
- L'article nécessite un contrôle de la température pour le stockage et la livraison avec des frais supplémentaires. Il n'est pas éligible au retour pour des raisons de sécurité et de qualité. Tenez compte des exigences avant d'acheter.
- Return Policy
Product Details & Documents
Stem cell factor (SCF) is a cytokine made by fibroblasts and endothelial cells. SCF binds to the receptor c-Kit/CD117 and plays a critical role in the maintenance, survival, and differentiation of hematopoietic stem cells. While human SCF shows no activity on murine cells, murine and rat SCF are active on human cells.
- High Quality
- Low Endotoxin
- Affordable
A COA is supplied with each product shipped detailing purity, formulation, biological activity, endotoxin levels, reconstitution instructions and recommended storage conditions. This product is for research use only and is not for use in humans.
Caution: For research use only.
Specifications for Product Family
Spécifications
Spécifications
- Catalog No:
- 10787-074
- 10787-070
- 10787-068
- 10822-360
- 10787-072
- Protein/peptide name:
- SCF
- SCF
- SCF
- SCF
- SCF
- Protein synonyms:
- c-Kit Ligand|KL|MGF|Steel Factor
- c-Kit Ligand|KL|MGF|Steel Factor
- c-Kit Ligand|KL|MGF|Steel Factor
- c-Kit Ligand|KL|MGF|Steel Factor
- c-Kit Ligand|KL|MGF|Steel Factor
- Protein/peptide type:
- Recombinant
- Recombinant
- Recombinant
- Recombinant
- Recombinant
- Protein Function:
- Cytokine
- Cytokine
- Cytokine
- Cytokine
- Cytokine
- Species:
- Human
- Human
- Human
- Human
- Human
- Source:
- E. coli
- E. coli
- E. coli
- E. coli
- E. coli
- Conjugation:
- Unconjugated
- Unconjugated
- Unconjugated
- Unconjugated
- Unconjugated
- Gene ID:
- P21583
- P21583
- P21583
- P21583
- P21583
- Sequence:
- MEGICRNRVT NNVKDVTKLV ANLPKDYMIT LKYVPGMDVL PSHCWISEMV VQLSDSLTDL LDKFSNISEG LSNYSIIDKL VNIVDDLVEC VKENSSKDLK KSFKSPEPRL FTPEEFFRIF NRSIDAFKDF VVASETSDCV VSSTLSPEKD SRVSVTKPFM LPPVA
- MEGICRNRVT NNVKDVTKLV ANLPKDYMIT LKYVPGMDVL PSHCWISEMV VQLSDSLTDL LDKFSNISEG LSNYSIIDKL VNIVDDLVEC VKENSSKDLK KSFKSPEPRL FTPEEFFRIF NRSIDAFKDF VVASETSDCV VSSTLSPEKD SRVSVTKPFM LPPVA
- MEGICRNRVT NNVKDVTKLV ANLPKDYMIT LKYVPGMDVL PSHCWISEMV VQLSDSLTDL LDKFSNISEG LSNYSIIDKL VNIVDDLVEC VKENSSKDLK KSFKSPEPRL FTPEEFFRIF NRSIDAFKDF VVASETSDCV VSSTLSPEKD SRVSVTKPFM LPPVA
- MEGICRNRVT NNVKDVTKLV ANLPKDYMIT LKYVPGMDVL PSHCWISEMV VQLSDSLTDL LDKFSNISEG LSNYSIIDKL VNIVDDLVEC VKENSSKDLK KSFKSPEPRL FTPEEFFRIF NRSIDAFKDF VVASETSDCV VSSTLSPEKD SRVSVTKPFM LPPVA
- MEGICRNRVT NNVKDVTKLV ANLPKDYMIT LKYVPGMDVL PSHCWISEMV VQLSDSLTDL LDKFSNISEG LSNYSIIDKL VNIVDDLVEC VKENSSKDLK KSFKSPEPRL FTPEEFFRIF NRSIDAFKDF VVASETSDCV VSSTLSPEKD SRVSVTKPFM LPPVA
- Protease-free:
- N
- N
- N
- N
- N
- Animal-free:
- N
- N
- N
- N
- N
- Nuclease-free:
- N
- N
- N
- N
- N
- Carrier-free:
- Y
- Y
- Y
- Y
- Y
- Endotoxin-free:
- N
- N
- N
- N
- N
- Endotoxin level:
- Low
- Low
- Low
- Low
- Low
- Endotoxin content:
- Endotoxin LAL (EU/µg) ≤0.1
- Endotoxin LAL (EU/µg) ≤0.1
- Endotoxin LAL (EU/µg) ≤0.1
- Endotoxin LAL (EU/µg) ≤0.1
- Endotoxin LAL (EU/µg) ≤0.1
- Biological activity:
- Bioactivity assay: TF-1 cell proliferation, ED50 ≤15 ng/ml, Bioactivity ≥6.7×10⁴ units/mg
- Bioactivity assay: TF-1 cell proliferation, ED50 ≤15 ng/ml, Bioactivity ≥6.7×10⁴ units/mg
- Bioactivity assay: TF-1 cell proliferation, ED50 ≤15 ng/ml, Bioactivity ≥6.7×10⁴ units/mg
- Bioactivity assay: TF-1 cell proliferation, ED50 ≤15 ng/ml, Bioactivity ≥6.7×10⁴ units/mg
- Bioactivity assay: TF-1 cell proliferation, ED50 ≤15 ng/ml, Bioactivity ≥6.7×10⁴ units/mg
- Purity:
- ≥95%
- ≥95%
- ≥95%
- ≥95%
- ≥95%
- Grade:
- RUO
- RUO
- RUO
- RUO
- Formulation:
- 10 mM sodium phosphate, 50 mM sodium chloride, pH 7.5
- 10 mM sodium phosphate, 50 mM sodium chloride, pH 7.5
- 10 mM sodium phosphate, 50 mM sodium chloride, pH 7.5
- 10 mM sodium phosphate, 50 mM sodium chloride, pH 7.5
- 10 mM sodium phosphate, 50 mM sodium chloride, pH 7.5
- Molecular weight:
- 18.6 kDa
- 18.6 kDa
- 18.6 kDa
- 18.6 kDa
- 18.6 kDa
- Reconstitution instructions:
- Sterile water at 0.1 mg/ml
- Sterile water at 0.1 mg/ml
- Sterile water at 0.1 mg/ml
- Sterile water at 0.1 mg/ml
- Sterile water at 0.1 mg/ml
- Storage conditions:
- –20 °C
- –20 °C
- –20 °C
- –20 °C
- –20 °C
- Shipping temperature:
- RT
- RT
- RT
- RT
- RT
- Tested applications:
- Bioassay
- Bioassay
- Bioassay
- Bioassay
- Bioassay
Spécifications
Product Family Options
Informations sur le produit
- Type de conditionnementTailleOriginAvailabilityPrice
- Vial2 μgUSDiscontinued-Supplier Number: -Spécifications:
More
Specifications
Cat. No.10787-074Protein/peptide nameSCFProtein synonymsc-Kit Ligand|KL|MGF|Steel FactorProtein/peptide typeRecombinantProtein FunctionCytokineSpeciesHumanSourceE. coliConjugationUnconjugatedGene IDP21583SequenceMEGICRNRVT NNVKDVTKLV ANLPKDYMIT LKYVPGMDVL PSHCWISEMV VQLSDSLTDL LDKFSNISEG LSNYSIIDKL VNIVDDLVEC VKENSSKDLK KSFKSPEPRL FTPEEFFRIF NRSIDAFKDF VVASETSDCV VSSTLSPEKD SRVSVTKPFM LPPVAProtease-freeNAnimal-freeNNuclease-freeNCarrier-freeYEndotoxin-freeNEndotoxin levelLowEndotoxin contentEndotoxin LAL (EU/µg) ≤0.1Biological activityBioactivity assay: TF-1 cell proliferation, ED50 ≤15 ng/ml, Bioactivity ≥6.7×10⁴ units/mgPurity≥95%GradeFormulation10 mM sodium phosphate, 50 mM sodium chloride, pH 7.5Molecular weight18.6 kDaReconstitution instructionsSterile water at 0.1 mg/mlStorage conditions–20 °CShipping temperatureRTTested applicationsBioassay - Supplier Number: 100-04-10UGSpécifications:
More
Specifications
Cat. No.10787-070Protein/peptide nameSCFProtein synonymsc-Kit Ligand|KL|MGF|Steel FactorProtein/peptide typeRecombinantProtein FunctionCytokineSpeciesHumanSourceE. coliConjugationUnconjugatedGene IDP21583SequenceMEGICRNRVT NNVKDVTKLV ANLPKDYMIT LKYVPGMDVL PSHCWISEMV VQLSDSLTDL LDKFSNISEG LSNYSIIDKL VNIVDDLVEC VKENSSKDLK KSFKSPEPRL FTPEEFFRIF NRSIDAFKDF VVASETSDCV VSSTLSPEKD SRVSVTKPFM LPPVAProtease-freeNAnimal-freeNNuclease-freeNCarrier-freeYEndotoxin-freeNEndotoxin levelLowEndotoxin contentEndotoxin LAL (EU/µg) ≤0.1Biological activityBioactivity assay: TF-1 cell proliferation, ED50 ≤15 ng/ml, Bioactivity ≥6.7×10⁴ units/mgPurity≥95%GradeRUOFormulation10 mM sodium phosphate, 50 mM sodium chloride, pH 7.5Molecular weight18.6 kDaReconstitution instructionsSterile water at 0.1 mg/mlStorage conditions–20 °CShipping temperatureRTTested applicationsBioassay - Supplier Number: 100-04-100UGSpécifications:
More
Specifications
Cat. No.10787-068Protein/peptide nameSCFProtein synonymsc-Kit Ligand|KL|MGF|Steel FactorProtein/peptide typeRecombinantProtein FunctionCytokineSpeciesHumanSourceE. coliConjugationUnconjugatedGene IDP21583SequenceMEGICRNRVT NNVKDVTKLV ANLPKDYMIT LKYVPGMDVL PSHCWISEMV VQLSDSLTDL LDKFSNISEG LSNYSIIDKL VNIVDDLVEC VKENSSKDLK KSFKSPEPRL FTPEEFFRIF NRSIDAFKDF VVASETSDCV VSSTLSPEKD SRVSVTKPFM LPPVAProtease-freeNAnimal-freeNNuclease-freeNCarrier-freeYEndotoxin-freeNEndotoxin levelLowEndotoxin contentEndotoxin LAL (EU/µg) ≤0.1Biological activityBioactivity assay: TF-1 cell proliferation, ED50 ≤15 ng/ml, Bioactivity ≥6.7×10⁴ units/mgPurity≥95%GradeRUOFormulation10 mM sodium phosphate, 50 mM sodium chloride, pH 7.5Molecular weight18.6 kDaReconstitution instructionsSterile water at 0.1 mg/mlStorage conditions–20 °CShipping temperatureRTTested applicationsBioassay - Supplier Number: 100-04-500UGL'article nécessite un contrôle de la température pour le stockage et la livraison avec des frais supplémentaires. Il n'est pas éligible au retour pour des raisons de sécurité et de qualité. Tenez compte des exigences avant d'acheter.Spécifications:
More
Specifications
Cat. No.10822-360Protein/peptide nameSCFProtein synonymsc-Kit Ligand|KL|MGF|Steel FactorProtein/peptide typeRecombinantProtein FunctionCytokineSpeciesHumanSourceE. coliConjugationUnconjugatedGene IDP21583SequenceMEGICRNRVT NNVKDVTKLV ANLPKDYMIT LKYVPGMDVL PSHCWISEMV VQLSDSLTDL LDKFSNISEG LSNYSIIDKL VNIVDDLVEC VKENSSKDLK KSFKSPEPRL FTPEEFFRIF NRSIDAFKDF VVASETSDCV VSSTLSPEKD SRVSVTKPFM LPPVAProtease-freeNAnimal-freeNNuclease-freeNCarrier-freeYEndotoxin-freeNEndotoxin levelLowEndotoxin contentEndotoxin LAL (EU/µg) ≤0.1Biological activityBioactivity assay: TF-1 cell proliferation, ED50 ≤15 ng/ml, Bioactivity ≥6.7×10⁴ units/mgPurity≥95%GradeRUOFormulation10 mM sodium phosphate, 50 mM sodium chloride, pH 7.5Molecular weight18.6 kDaReconstitution instructionsSterile water at 0.1 mg/mlStorage conditions–20 °CShipping temperatureRTTested applicationsBioassay - Supplier Number: 100-04-1MGSpécifications:
More
Specifications
Cat. No.10787-072Protein/peptide nameSCFProtein synonymsc-Kit Ligand|KL|MGF|Steel FactorProtein/peptide typeRecombinantProtein FunctionCytokineSpeciesHumanSourceE. coliConjugationUnconjugatedGene IDP21583SequenceMEGICRNRVT NNVKDVTKLV ANLPKDYMIT LKYVPGMDVL PSHCWISEMV VQLSDSLTDL LDKFSNISEG LSNYSIIDKL VNIVDDLVEC VKENSSKDLK KSFKSPEPRL FTPEEFFRIF NRSIDAFKDF VVASETSDCV VSSTLSPEKD SRVSVTKPFM LPPVAProtease-freeNAnimal-freeNNuclease-freeNCarrier-freeYEndotoxin-freeNEndotoxin levelLowEndotoxin contentEndotoxin LAL (EU/µg) ≤0.1Biological activityBioactivity assay: TF-1 cell proliferation, ED50 ≤15 ng/ml, Bioactivity ≥6.7×10⁴ units/mgPurity≥95%GradeRUOFormulation10 mM sodium phosphate, 50 mM sodium chloride, pH 7.5Molecular weight18.6 kDaReconstitution instructionsSterile water at 0.1 mg/mlStorage conditions–20 °CShipping temperatureRTTested applicationsBioassay