ContactUsLinkComponent
Energy Transfer Apparatus
Catalogue # 470163-046
Fournisseur:  Wards
undefined
Energy Transfer Apparatus
Catalogue # 470163-046
Fournisseur:  Wards
Supplier Number:  PH0467B
Spécifications
  • Description:
    Energy Transfer Apparatus
  • Cat. no.:
    470163-046
  • Supplier No.:
    PH0467B
Spécifications
À propos de cet article
Demonstrate the conversion from electrical energy to mechanical energy
  • Investigate electrical to mechanical energy
Demonstrate the conversion of energy with this small motor and its' attached pulley. The unit can also be used with a larger motor/generator to compare power input and output. Drive each in turn with the other while metering the voltage and current.
Motor generator and hanging masses sold separately.