VWR® Flat-Bottom Boiling Flasks
VWR® Flat-Bottom Boiling Flasks
Supplier:  VWR International
Certificates
VWR® Flat-Bottom Boiling Flasks
VWR® Flat-Bottom Boiling Flasks
2 Options Available Below
Product Details & Documents
The flasks come with uniform wall thickness which makes these flasks ideal for heating.
  • Borosilicate glass fabrication
  • Good chemical resistance
Durable product has good temperature and chemical resistance.
Manufactured from ASTM E438 Type 1 Class A borosilicate 33 expansion glass. As per ASTM specification E 1403, Type 1 Class 4.
Product Family Options
Product Information
  • Capacité
    Neck Size
    Availability
    Price
  • 300 ml
    24/40
    Log in for availability
    Log in for availability
    Log in for availability
    Pack of 2
    Pack of 2 - $133.17
    $133.17
    New Customer Promotional Price
    Log in for availability
    Case of 12
    Case of 12 - $799.01
    $799.01
    New Customer Promotional Price
    Pack of 2
       $133.17
    Case of 12
       $799.01
  • 500 ml
    24/40
    Log in for availability
    Log in for availability
    Log in for availability
    Pack of 2
    Pack of 2 - $93.16
    $93.16
    New Customer Promotional Price
    Log in for availability
    Case of 12
    Case of 12 - $558.96
    $558.96
    New Customer Promotional Price
    Pack of 2
       $93.16
    Case of 12
       $558.96