Worm Gearbox
Worm Gearbox (216.1:1 & 336.1:1)
Catalog # 470096-272
Supplier: TAMIYA AMERICA INC.
Fournisseur #: 72004
Jump to:
Product Details & Documents
With the Worm Gear Box, High Efficiency, it is possible to have high power or high speed with two different gear ratios
Several crank arms have been supplied along with a RE-260 motor.
Spécifications
- Désignation:Worm Gearbox (216.1:1 & 336.1:1)