High Speed Gearbox | Avantor
High Speed Gearbox
High Speed gearbox(11.6:1 & 18:1)
Catalog # 470096-268
Supplier:   TAMIYA AMERICA INC.
Fournisseur #:  72002
High Speed Gearbox
Catalog # 470096-268
High Speed Gearbox
High Speed Gearbox
High Speed gearbox(11.6:1 & 18:1)
Price
$35.50
Log in pour la disponibilité
Each
Product Details & Documents
The High Speed Gear Box, High Efficiency, is perfect for high speed applications
Two gear ratios are possible, and several styles of crank arms are supplied for a variety of uses. A RE-260 motor is included. The gear box can be mounted vertically or horizontally.
Spécifications
  • Désignation:
    High Speed gearbox(11.6:1 & 18:1)