Quantofix Lubricheck Test Strips, CTL Scientific Supply
Catalogue # 10217-568
Fournisseur: Macherey-Nagel
Spécifications
- Description:Lubricheck Test Strips
- Storage:RT
- Cat. no.:10217-568
- Scale:0-15-50-75-130-200 mmol/L KOH
- Supplier No.:91336
Spécifications
À propos de cet article
Quantofix Lubricheck test strip for the semi-quantitative determination of the concentration of cooling lubricants
Quantofix Lubricheck has a gradation of scale: 0-15-50-75-130-200 mmol/L KOH.
The easy dip and read procedure provides a reliable result within 1 minute.
Ordering information: Packaged as a box of 100.