Quantofix Silver Test Strips, CTL Scientific Supply
Silver test strips, 0 - 1 - 2 - 3 - 5 - 7 - 10 g/L
Supplier: Macherey-Nagel
Fournisseur #: 91350
Quantofix Silver Test Strips, CTL Scientific Supply
Catalog # 10217-556
Supplier: Macherey-Nagel
CAS Number:
Jump to:
Product Details & Documents
Quantofix Silver test strips for the semi-quantitative determinaton of silver.
Quantofix Silver has a gradation of scale: 0-1-2-3-5-7-10.
Ordering information: Packaged as a box of 100.
Spécifications
Spécifications
- Désignation:Silver test strips
- Range:0 - 1 - 2 - 3 - 5 - 7 - 10 g/L
- Cat. No.:10217-556
- Storage:RT