Plastic Beaker Set, PMP
50 / 100 / 250 / 500 / 1000 ml, Plastic Beaker Set, PMP, Set of 5
Catalog # 470164-038
Supplier: United Scientific Supplies
Fournisseur #: BMSET5
Jump to:
Product Details & Documents
This durable set of beakers includes five different sizes.
- Graduated Markings
- Five Different Sizes
- Polymethylpentene (PMP) Construction
- Durable
- Clear Walls
Made of polymethylpentene, the beakers offer glass-like clarity with the durability of plastic.
All beakers in the set have graduated markings.
Ordering information: This set includes five graduated PMP beakers in sizes 50 mL, 100 mL, 250 mL, 500 mL, and 1000 mL.
Spécifications
- Capacité:50 / 100 / 250 / 500 / 1000 ml
- Désignation:Plastic Beaker Set, PMP, Set of 5