CENCO® Assorted Ball Set
CENCO® Assorted Ball Set
Catalog # 470164-716
Supplier: United Scientific Supplies
Fournisseur #: DBLST12-A
Jump to:
Product Details & Documents
Comprehensive and High Quality.
- Packed in plastic storage box
- Set of 12
A set with 12 balls in all, includes 25mm diameter drilled balls of brass, aluminum, wood, steel, copper and lead; two 25mm solid steel balls, one plain and one with bifilar suspension lugs; 38 mm balls of brass, steel and lead with hooks; and one 25mm solid cork ball.
Spécifications
- Désignation:CENCO® Assorted Ball Set