Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
- Description:Specimen Chuck Stand
- Dimensions:8 7/8 x 2 3/4 x 1 1/4"
- Cat. no.:10764-046
- Supplier No.:SDST001
This stainless steel stand is used to store chucks both outside and inside the cooling chamber.
This small metal stand fits five specimen chucks up to 55 mm in diameter. Specimen chucks are not included. High quality stainless steel stand is perfect for in-chamber and can be used to expand the internal chuck capacity of the cryostat.