Precellys® CK68 Lysing Kit, Dry Hard Tissue Grinding, Bertin
CK68-R is made with inert 2.8 mm ceramic (zirconium oxide) beads.
- Autoclavable
- Usable with Precellys®24, Precellys® Evolution and Minilys® homogenizers
It is designed for dry samples grinding like rice grains, pepper grains and more.
Frequently Bought Together