À propos de cet article
Whatever you cleaning requirements, UltraSpeed Pro will dramatically raise your standards in speed, performance, versatility and ergonomics
The UltraSpeed and UltraSpeed Pro Systems both feature a lightweight, durable autoclavable frame. The 16" frame can be used with Ultraspeed pads, standard pocket mops and flat sponge cleanroom mops. Easy, fast rinsing and wringing of mops without removal of the frame. Improved foot pedal is larger and easier to step on and helps improve insertion into the wringer.
The UltraSpeed Pro range includes options to suit all sites, starting with the single bucket system for general floor cleaning. Add a second bucket onto the chassis and you have a double bucket system for mopping larger areas. If you want to fulfill more cleaning tasks you can use the second bucket to carry wipes, chemicals, etc. Each solution is available with an ergonomic push bar that makes moving the system around sites simple and can be used with your choice of microfiber mops as well as specialty mops for cleaning safety floors.
Compared to current bucket-and-press systems, the UltraSpeed Pro flat mopping system outperforms them in every aspect. Kit contains: telescopic handle, bucket, chassis, press, mop frame and mop pad.