À propos de cet article
The CKMix50-R and CKMix50 Tissue Grinding Kits are designed for hard tissue grinding and homogenizing
- Dnase/Rnase free
- Inert 2.8 mm and 5.0 mm ceramic (zirconium oxide) beads
- Autoclavable
- Ideal for hard tissue grinding
The CKMix50 Tissue Grinding Kit contains 7 mL standard tubes, and is ideal for use with whole organs such as rat brain, liver, kidney, skin, whole iris-ciliary body pork, and dry samples such as pepper grains. All kits include 50 preps, ready-to-use with the Precellys®24, Precellys® Evolution, or Minilys devices. All tubes are manufactured from a polypropylene copolymer, and are resistant to all organic solvents.
Kits feature prefilled 2.8 mm and 5.0 vmm ceramic (zirconium oxide) beads in tubes. The CKMix50-R Tissue Grinding Kit contains 2 mL custom designed reinforced tubes that are intended for bead beating applications, with O-Rings and reinforced materials for dry grinding valuable or very hard samples. The CKMix50-R kit is ideal for use with hair, mouse lung, mouse heart, dry mouse hair, and mouse bone samples