Precellys® VK05 Lysing Kit, Tough Micro-Organism, Bertin
Supplier: BERTIN CORP
Certificates
About this item
The VK05 lysing kit is designed for tough micro-organism lysing like yeast, fungi, cyanobacteria, phytoplankton, nematode, spores, micro-algae and more.
- Inert 0.5 mm glass beads
- Autoclavable
- Usable with Precellys®24, Precellys® Evolution and Minilys® homogenizers
Frequently Bought Together