Precellys® CK28 Lysing Kit, Hard Tissue Homogenizing, Bertin
Supplier: BERTIN CORP
Certificates
About this item
CK28 is made with inert 2.8 ceramic (zirconium oxide) beads.
- Autoclavable
- Usable with Precellys®24, Precellys® Evolution and Minilys® homogenizers
It designed for hard tissue homogenization like ear, lung, heart, spleen, tail, artery, arabidopsis, leaves and more.
Frequently Bought Together