Minilys Homogenizer, Bertin
Catalog # 10144-650
Supplier: BERTIN CORP
Specifications
- Capacity:Up to 3 tubes of 0.5, 1.5, or 2 mL
1 tube of 7 mL - Description:Minilys Homogenizer
- Dimensions:180L x 290W x 380H mm
- Sound level:< 70 dB
- No. of cycles:1 Cycle (up to 240 sec.)
- Weight:7.8 kg (17.2 lbs.)
- Cat. no.:10144-650
- Electrical:100/230 VAC, 50/60 Hz, 1 kVA
- Supplier No.:06404200-RD000
Specifications
About this item
The Minilys is a compact and efficient homogenizer dedicated to sample preparation
- 3 speed settings: 3000, 4000 and 5000 rpm
- Manual tube locking system
- Capacity: three tubes of 0.5, 1.5 or 2mL and one tube of 7mL
- Simple user interface with two parameters (speed and time)
The unit allows users to homogenize, grind, or lyse any type of sample in as little as 30 seconds, due to 3D motion movement. The Minilys Homogenizer is ideally suited for daily use in sample homogenization.
Certifications: CE certified. FCC part 15 certified.