Precellys® CK14 Lysing Kit, Soft Tissue Homogenizing, Bertin
Precellys® CK14 Lysing Kit, Soft Tissue Homogenizing, Bertin
   Sustainable Options Available
Supplier:   BERTIN CORP
Precellys® CK14 Lysing Kit, Soft Tissue Homogenizing, Bertin
Supplier:  BERTIN CORP
   Sustainable Options Available
Certificates
Precellys® CK14 Lysing Kit, Soft Tissue Homogenizing, Bertin
Precellys® CK14 Lysing Kit, Soft Tissue Homogenizing, Bertin
4 Options Available Below
Product Details & Documents
CK14 is made with inert 1.4 mm ceramic (zirconium oxide) beads.
It is designed for soft tissue homogenization like brain, liver, kidney, skin, plant leaves, mammalian cells and more.
Product Family Options
Product Information
  • Désignation
    Taille
    Volume
    Availability
    Price
  • CK14 soft tissue homogenizing kit, tubes with screw cap and skirt
    20 – 200 mg, or 200 µL to 1.6 mL
    2 ml tubes
    Log in for availability
    Log in for availability
    Each
    Each - $421.17
    $421.17
    Each
       $421.17
    Supplier Number: P000912-LYSK1-A.0
    Specifications:
    More
    Specifications
    Cat. No.
    10144-496
  • CK14 soft tissue homogenizing kit, tubes with screw cap, O-ring, and skirt
    1 – 20 mg, or 5 µL to 200 µL
    0.5 ml tubes
    Log in for availability
    Log in for availability
    Each
    Each - $421.17
    $421.17
    Each
       $421.17
    Supplier Number: P000933-LYSK0-A.0
    Specifications:
    More
    Specifications
    Cat. No.
    10144-498
  • CK14 soft tissue homogenizing kit, tubes with screw cap, O-Ring, and skirt
    200 mg to 2 g, or 200 µL to 6 mL
    7 ml tubes
    Log in for availability
    Log in for availability
    Each
    Each - $602.46
    $602.46
    Each
       $602.46
    Supplier Number: P000940-LYSK0-A.0
    Specifications:
    More
    Specifications
    Cat. No.
    10144-536
  • CK14 Soft Tissue Homogenizing Kit, beads in bulk format, 325 g of inert 1.4 mm ceramic beads, beads only
    Log in for availability
    Log in for availability
    Each
    Each - $454.36
    $454.36
    Each
       $454.36
    Supplier Number: P000927-LYSK0-A.0
    Specifications:
    More
    Specifications
    Cat. No.
    10144-554