Anti-beta Actin Mouse Monoclonal Antibody [clone: 2D1D10] (HRP (Horseradish Peroxidase))
THE™ beta Actin Antibody [HRP], mAb, Mouse reacts with mouse, rabbit, chicken, human, hamster, cow, goat, fish, and pig substrates.It has not yet been tested in other species.
- High specificit
- High affinity
- High stability
GenScript offers a wide selection of polyclonal and monoclonal loading control antibodies for α-tubulin and β-actin for several species. HRP- and biotin-conjugated versions of these antibodies are also available. GenScript also offers a series of isotype control antibodies that can be used in your research.
Type: Tag
Antigen: ACTB
Clonality: Monoclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity:
- Catalog No:
- 89493-630
- 76198-724
- Antigen symbol:
- beta Actin
- beta Actin
- Antigen name:
- beta Actin
- beta Actin
- Antigen synonyms:
- PS1TP5-binding protein 1|cytoplasmic 1|PS1TP5BP1|BRWS1|beta cytoskeletal actin|actin
- Antibody type:
- Control
- Control
- Clonality:
- Monoclonal
- Monoclonal
- Conjugation:
- HRP (Horseradish Peroxidase)
- HRP (Horseradish Peroxidase)
- Clone:
- 2D1D10
- 2D1D10
- Host:
- Mouse
- Mouse
- Isotype:
- IgG2a
- IgG2a
- Western blot:
- Yes
- Yes
- ELISA:
- Yes
- Yes
- Immunogen:
- A synthetic peptide DDDIAALVVDNGSG coupled to KLH
- A synthetic peptide DDDIAALVVDNGSG coupled to KLH
- Form:
- Lyophilized
- Lyophilized
- Purification:
- Protein G affinity column
- Protein G affinity column
- Concentration:
- Reconstitute the lyophilized powder with deionized water (or equivalent) to an final concentration of 0.5 mg/ml
- Reconstitute the lyophilized powder with deionized water (or equivalent) to a final concentration of 0.5 mg/ml
- Cross adsorption:
- No
- No
- Storage buffer:
- PBS, pH 7.4, containing 1% BSA and 0.01% thimerosal
- PBS, pH 7.4, containing 1% BSA and 0.01% thimerosal
- Storage temperature:
- The antibody is stable in lyophilized form if stored at -20 °C or below. The reconstituted antibody can be stored for 2-3 weeks at 2...8 °C. For long term storage, aliquot and store at -20 °C or below. Avoid repeated freezing and thawing cycles.
- The antibody is stable in lyophilized form if stored at −20 °C or below. The reconstituted antibody can be stored for 2 - 3 weeks at 2 - 8 °C. For long term storage, aliquot and store at −20 °C or below. Avoid repeated freezing and thawing cycles
- Shipping temperature:
- Ice
- Ice
Frequently Bought Together