About this item
Bio-Rad offers acrylamide, bis-acrylamide, and premixed acrylamide/bis-acrylamide
- Acrylamide is available as a powder or 40% solution
- Bis-acrylamide is available as a powder or 2% solution
- Premixed acrylamide/bis solutions are available as either 30% or 40% acrylamide with acrylamide:bis-acrylamide ratios of 19:1, 29:1 or 37.5:1
- Premixed acrylamide/bis powders are available with acrylamide:bis-acrylamide ratios of 19:1, 29:1 or 37.5:1
- TGX™ FastCast™ and TGX Stain-Free™ FactCast™ Kits (with acrylamide solutions and buffers) are available for 7.5%, 10%, or 12% acrylamide gels
TGX™ FastCast™ and TGX Stain-Free™ FactCast™ premixed acrylamide solutions and kits use Tris-glycine eXtended chemistry. Kits have a one year shelf life, and cast gels can be stored for up to a month. TGX™ Gels can separate proteins in only 20 minutes and have Laemmli-like separation characteristics when using a standard Tris/glycine/SDS running buffer system. Stain-free technology permits visualization of proteins in a gel or blot in less than 5 minutes with stain-free enabled imaging systems (Bio-Rad’s Gel Doc™ EZ, Gel Doc™ XR+, ChemiDoc™ MP, and ChemiDoc™ XRS+ Imaging Systems).
All are available as both powders and solutions.
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.