ContactUsLinkComponent
Set of Casters for 6 Gallon Buckets, Vileda Professional®
Set of Casters for 6 Gallon Buckets, Vileda Professional®
Catalog # 10153-934
undefined
Set of Casters for 6 Gallon Buckets, Vileda Professional®
Catalog # 10153-934
Supplier Number:  135083
Specifications
  • Description:
    Casters for 22.7 L (6 gal.) buckets
  • Cat. no.:
    10153-934
  • Supplier No.:
    135083
Specifications
About this item
Casters for six gallon buckets that are ideal for 10 and 14" Roll-O-Matic® mops.
  • Fits securely into six gallon buckets
  • Easily snaps into place
  • Not autoclavable
Casters sold separately from buckets.