Certified WebSeal™ Plates, Plastic, Non-Coated, Nonsterile, Thermo Scientific
Supplier: Thermo Scientific
Certificates
About this item
Manufactured from ultra-low bleed, high-purity basis resin
Fully lot tested by GC-MS and LC/MS for organic extractables.
Frequently Bought Together