Face Shield, Unimed-Midwest
Catalog # 89068-863
Supplier: Unimed-Midwest
Specifications
- Description:Full Face Shield
- Size:13"W x 7.5" L
- Cat. no.:89068-863
- Supplier No.:SSDS00205B
Specifications
About this item
Splash shield provides comfortable face protection.
- Soft elastic strap wraps around head to secure shield
- Anti-fog
- Optically Clear
- 13 inch impenetrable foam barrier for wide coverage
- Disposable and single use
It is not recommended to disinfect and reuse shields because doing so would destroy the anti-fog coating.