Carts, Element Series, TRIPPNT
Carts, Element Series, TRIPPNT
Supplier:   TrippNT
Carts, Element Series, TRIPPNT
Supplier:  TrippNT
Certificates
Carts, Element Series, TRIPPNT
Carts, Element Series, TRIPPNT
4 Options Available Below
Product Details & Documents
TrippNT's Element Cart Series is constructed of 2 skins of corrosion resistant aluminum with a high density polyethylene core
  • Fully customizable corrosion resistant carts using built in tracks
  • Superior maneuverability
  • Constructed with polyethylene and aluminum materials
  • Rugged, flexible, movable, workspace and storage
  • Delivered fully assembled, no tools required
  • Made in the USA
  • 100% Total Replacement Lifetime Guarantee
Order a base model and then add whatever accessories fit your work situation. All TrippNT Carts have a 100% Total Replacement Lifetime Guarantee. White color. Polyethylene and Aluminum material.
Each cart is fully customizable using built in tracks and has superior maneuverability and 4-inch nylon rubber non-marking casters.
Accessories information: Accessories can be added to any cart in the series. Add IV poles, sharps containers, trash cans, gas tanks and cardiac boards. Rugged, flexible, movable, workspace and storage. Delivered fully assembled, no tools required. Accessories can also be ordered separately to add to previously purchased carts. Made in the USA. 100% total replacement lifetime guarantee.
Product Family Options
Informations sur le produit
  • Désignation
    Color
    Largeur
    Hauteur
    Profondeur
    No. of Drawers
    Weight
    Availability
    Price
  • Element 02, Procedure Cart
    White
    32"
    43"
    23"
    6
    88 lbs.
    Discontinued
    Supplier Number: -
  • Element 01, Locking Cart, 6 Side Shelves
    White
    24"
    36"
    19"
    3
    62 lbs.
    Discontinued
    Supplier Number: -
  • Element 03, Space Saving Cart
    White
    21"
    35.5"
    18.5"
    3
    57 lbs.
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $5,194.81
    $5,194.81
    Each
       $5,194.81
    Minimum order quantity is 1
    Supplier Number: 51498
  • Element 04, Extra Tall Tower Cart
    White
    24"
    47"
    18"
    6
    77 lbs.
    Discontinued
    Supplier Number: -
  • Element 05, with Railtop Standard, All Purpose Cart, 6 Side Shelves
    White
    26.5"
    43"
    21"
    3
    62 lbs.
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $9,272.62
    $9,272.62
    Each
       $9,272.62
    Minimum order quantity is 1
    Supplier Number: 51504
  • Element 06, with Railtop Standard, Tall Space Saving Cart
    White
    26.5"
    43"
    20"
    3
    47 lbs.
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $5,455.87
    $5,455.87
    Each
       $5,455.87
    Minimum order quantity is 1
    Supplier Number: 51609
  • Element 07, Compact Cart
    White
    17"
    17"
    27"
    4
    35 lbs.
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $4,628.28
    $4,628.28
    Each
       $4,628.28
    Minimum order quantity is 1
    Supplier Number: 51610
  • Discontinued
    Supplier Number: -
Accessories
Informations sur le produit
  • Catalog #
    For Use With
    Availability
    Price
  • Cardiac Board and Mount
    Cardiac Board and Mount
    10020-700
    Elements: 02,02C,09
    Log in pour la disponibilité
    Each - $372.80
Informations sur le produit
  • Catalog #
    For Use With
    Availability
    Price
  • Electronic Lock
    Electronic Lock
    10020-706
    Elements: 01,02,02C,03,04,05,06,07,09
    Log in pour la disponibilité
    Each - $544.78