VWR® PCR Workstations
Product Details & Documents
The VWR® PCR Workstation offers twin decontamination action by UV inactivation of airborne and surface-bound contaminants, and therefore represents an ideal environment for PCR sample preparation and other sensitive protocols.
- Large working area for stress-free working
- Can accommodate benchtop equipment
- Function indicator for UV air recirculator tube
- Displays operating time of UV tubes
- Enables well-timed replacement of tubes for constant UV intensity
- Removable shelves provide additional storage space at the rear panel
- Four internal power outlets for operating lab equipment
- Electromagnetic safety mechanism stops UV irradiation if front panel is opened
This workstation allows the combination of several working steps without change of location or interruption of workflow, thereby also minimising the risk of cross-contamination.
Active decontamination of work surface during non-working time by UV irradiation. Additional inactivation of aerosol-bound contaminants by shielded UV Air Recirculator during operation. Contaminant prevention with antimicrobial work surface made of stainless steel.
Ordering information: Assembly required.
Specifications
- Désignation:PCR Workstation
- Cat. No.:10783-132
- Working area:72L×54D cm
- Exterior dimensions:75L×78H×62D cm
Accessories
Product Information
- Catalog #AvailabilityPrice
- Replacement Interior White Light Tube, for UV Air Recirculator or HEPA/UV system, 15 W, pack of 1Replacement Interior White Light Tube, for UV Air Recirculator or HEPA/UV system, 15 W, pack of 110783-134Log in for availabilityEach - $48.60
Product Information
- Catalog #AvailabilityPrice
- Replacement Interior UV Tubes for VWR® PCR Workstation, 8 W (254 nm), pack of 2 tubesReplacement Interior UV Tubes for VWR® PCR Workstation, 8 W (254 nm), pack of 2 tubes10783-136Log in for availabilityEach - $152.86
Product Information
- Catalog #AvailabilityPrice
- Replacement Interior UV Tube for UV Air Recirculator or HEPA/UV system, 8 W (254 nm), pack of 1Replacement Interior UV Tube for UV Air Recirculator or HEPA/UV system, 8 W (254 nm), pack of 110783-138Log in for availabilityEach - $74.51
Product Information
- Catalog #AvailabilityPrice
- Replacement Dust Filter for VWR® PCR Workstation Air Recirculator, pack of 10 filters (shelf life >10 years if stored dry at room temperature)Replacement Dust Filter for VWR® PCR Workstation Air Recirculator, pack of 10 filters (shelf life >10 years if stored dry at room temperature)10783-140Log in for availabilityPack of 10 - $38.26