Weighing Dishes, Wheaton
30617-222
Small Business Enterprise
- Dimensions:Dia.×H 20×8 mm (⁴⁄₅׳⁄₁₀")
- Material:Aluminium
- Cat. no.:30617-222
- Supplier No.:370792
This weighing dishes are made from aluminum which is ideal for milligram weighing.
- For use with liquids or solids
- Smooth surface ensures complete transfer
- Use of balance-type forceps enables easy grasping